Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Liofilchem™ Contam Swab Salmonella
SDP

Catalog No. 22777006 Shop All Liofilchem Products
Change view
Click to view available options
Detection Method:
Rapid Detection
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detection Method
22-777-006 Rapid Detection
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-777-006 Supplier Liofilchem™ Supplier No. 86103
Add to Cart
Add to Cart

A microbiological test with a selective and differential medium for the detection of Salmonella spp. directly from surfaces.

  • Ready-to-use-sterile Rayon swab
  • Selective and differential medium
  • Kit contents: 30 sealed envelopes (swab + culture medium) and one instruction sheet
  • Watch for the color change of the medium and interpret the results

Specifications

Sample Type Environmental Sample
Detection Method Rapid Detection
For Use With (Application) Work Surface Contamination Monitoring
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.