Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Liofilchem™ Contam Swab Yeasts/Moulds
SDP

Catalog No. 22777014 Shop All Liofilchem Products
Change view
Click to view available options
Detection Method:
Rapid Detection
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detection Method
22-777-014 Rapid Detection
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-777-014 Supplier Liofilchem™ Supplier No. 86105
Add to Cart
Add to Cart

A swab containing a culture medium used for the detection of yeasts and moulds directly from surfaces.

  • The product consists of an envelope made of transparent paper/plastic bag containing a culture medium and a sterile swab
  • Detection of yeasts and moulds directly from surfaces
  • Kit contents: 30 sealed envelopes (swab + culture medium) and one instruction sheet
  • Watch for the color change of the medium and interpret the results

Specifications

Detection Method Rapid Detection
Sample Type Environmental Sample
For Use With (Application) Work Surface Contamination Monitoring
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.