Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contec™ Edgeless™ Mop System
BIOPHARMAGSA_VAGREENER_CHOICE

Catalog No. 19033950
Click to view available options
Head Type:
Edgeless
Enhanced with Textured Headband
Long Textured
Ribbed
Textured
Color:
Black (Tail Band)
Gray, Black (Tail Band)
White
White, Blue (Tail Band)
White, Red (Tail Band)
Quantity:
1/Bag, 20 Bags/Cs.
1/Bag, 30 Bags/Cs.
12/Cs.
20/Cs.
30/Cs.

Clean in controlled environments and cleanrooms. The Edgeless™ Mop System is constructed from a continuous tube of 100% polyester knitted fabric. The mop heads help eliminate contamination from breakage and deterioration of traditional string mops.

  • Designed for wet mopping in controlled environments and cleanrooms
  • Ideal for use in sterile environments
  • Mop heads are constructed from a continuous tube of 100% polyester knitted fabric, looped to form strands that encapsulate liquids for maximum absorption
  • Mop heads are free of any cellulosic contaminants
  • Textured mop heads feature ribbed, twisted strands for 60% more sorbent capacity than standard Edgeless mops
  • Textured knitted polyester tube mop heads help eliminate contamination from breakage and deterioration of traditional string mops
  • Mop heads are laundered and packaged in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Mop Head
Head Type Textured
Color White
Sterility Non-sterile
Autoclavable Autoclavable
Disposable Limited-use
Quantity 1/Bag, 30 Bags/Cs.
For Use With (Equipment) Contec Edgeless Mopping System
Description Cleanroom Laundered Textured Mop Head
For Use With (Application) Designed for use with aqueous solutions containing cleaning agents and disinfectants; compatible with wide range of chemicals
Material Polyester
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.