Learn More
Contec™ QuicKlean™ with MicroGenesis™ Mop and Wipe System
Designed for USP <797> compliance in healthcare cleaning. The Contec™ QuicKlean™ with MicroGenesis™ Mop and Wipe System combines lightweight, easy to maneuver hardware and the particle pick-up capacity of the MicroGenesis microfiber wipe to deliver unprecedented performance. It is compatible with a wide range of disinfectants, chemicals, and solutions.
Manufacturer:  Contec™ 2745
 View more versions of this product
Catalog No. 19-706-095
  Check Availability
Add to cart
Specifications
Description
  • Lightweight mophead or fram with MicroGenesis microfiber for USP and healthcare cleaning
  • Compatible with a wide range of disinfectants, chemicals and solutions
  • Clean and disinfect floors, wall and ceilings
  • Withstands daily use with little fatigue on user
  • Autoclavable hardware
  • Safe for use with all disinfectants including bleach and quaternary ammonium
  • Handle sold separately
Compliance
USP <797>
Description & Specifications
Specifications
Each
USP 797
Polypropylene
Ideal for disinfecting floors, walls and ceilings
16 in.
Blue
Yes
Flat mop head frame
5 in.
5 x 16 in. (13 x 41cm)