Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contec™ Sporicidin™ Antimicrobial Lotion Soap

Catalog No. 189991454 Shop All Contec Inc. Products
Click to view available options
Quantity:
16 oz.
1 gal.

Catalog No. 18-999-1454

Supplier: Contec™ CALS2416

Use daily for handwashing prior to donning gloves or beginning work. The Sporicidin™ Antimicrobial Lotion Soap is Ideal for use in ISO Class 6 and 7 Buffer Zones and ISO Class 8 anterooms in hospitals and compounding pharmacies. Contains no triclosan, PCMX or chlorhexidine.

  • Used daily for handwashing prior to donning gloves, or beginning work in isolators, hospitals and compounding pharmacies
  • Alcohol-free
  • Contains no triclosan, PCMX, or chlorhexidine
  • Kills up to 99.99% of disease- and odor-causing organisms FDA categorized as a “Health Care Personnel Handwash” and “Surgical Scrub”
  • Recommended for repeated daily use
  • Gentle and soothing to the skin
  • Soap is also an effective skin antiseptic and deodorant

Specifications

Certifications/Compliance ISO 6, 8
For Use With (Application) Used daily for handwashing prior to donning gloves, or beginning work in isolators, hospitals and compounding pharmacies
Quantity 16 oz.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.