Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Contec™ Sporicidin™ Antimicrobial Lotion Soap
Shop All Contec Inc. Products
Click to view available options
Quantity:
16 oz.
1 gal.
Description
- Used daily for handwashing prior to donning gloves, or beginning work in isolators, hospitals and compounding pharmacies
- Alcohol-free
- Contains no triclosan, PCMX, or chlorhexidine
- Kills up to 99.99% of disease- and odor-causing organisms FDA categorized as a “Health Care Personnel Handwash” and “Surgical Scrub”
- Recommended for repeated daily use
- Gentle and soothing to the skin
- Soap is also an effective skin antiseptic and deodorant
Specifications
Specifications
| Certifications/Compliance | ISO 6, 8 |
| For Use With (Application) | Used daily for handwashing prior to donning gloves, or beginning work in isolators, hospitals and compounding pharmacies |
| Quantity | 16 oz. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction