Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contec™ Sporicidin™ Disinfectant Solution, Nonsterile
BIOPHARMAGREENER_CHOICE

Catalog No. p-4164022
Click to view available options
Volume (Metric):
0.94 L
3.8 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Disinfects, cleans, and deodorizes hard surfaces. Contec™ Sporicidin™ Disinfectant Solution is compatible with plastic, wood, glass, and metal surfaces.

  • Provides 100% kill of pathogenic vegetative organisms, including MRSA, VRE and Avian Influenza A Virus (H9N2 and H1N1)
  • Continuous residual activity up to 6 months
  • Compatible with plastics, wood, glass and metals
  • Non-staining, non-abrasive and non-corrosive
  • Alcohol-free
  • EPA and FDA registered
  • OSHA compliant
  • Lowest EPA toxicity category

Compliance

EPA and FDA registered, OSHA compliant, Lowest EPA toxicity category

Specifications

Product Type Disinfectant Solution
Form Liquid
pH 7.5 to 8.5
Product Line Sporicidin™
Sterility Non-sterile
Certifications/Compliance EPA, OSHA
Composition 1.56% Buffered Phenol, 0.06% Sodium Phenate, 98.38% Inert Ingredients
For Use With (Application) Disinfecting hard surfaces in critical areas where USP <797> compliance has been mandated, such as ISO Class 8 Anterooms in hospitals and compounding pharmacies
Volume (English) 1 gal.
Volume (Metric) 3.8 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.