Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Contec™ Sporicidin™ Disinfectant Wipes

Shop All Contec Inc. Products


Click to view available options
Dimensions (L x W):
9.5 x 12 in.
5 x 8 in.
Description
- Provides 100% kill of pathogenic vegetative organisms, including MRSA, VRE and Pandemic 2009 H1N1 Influenza A Virus (as well as H9N2)
- Continuous residual activity up to 6 months after use
- Wipes are non-staining, non-abrasive and non-corrosive
- Recommended for use on treated wood, metal, painted surfaces, glass, porcelain, glazed tile, plastic, rubber, and vinyl materials
- Provides users with a product safety rated in the lowest EPA toxicity categories
- EPA and FDA registered; OSHA compliant
- Non-hazardous disinfectant solution has no special shipping or disposal restrictions
Compliance
EPA registered. OSHA compliant.
Specifications
Specifications
| Product Type | Disinfectant Wipe |
| Certifications/Compliance | EPA, OSHA |
| Dimensions (L x W) | 5 x 8 in. |
| For Use With (Application) | Wiping cabinets, laminar flow hoods and other surfaces, compatible with Plastics, Wood, Glass and Metal, hard surfaces in critical areas |
| Packaging Quantity | 180/Canister, 12 Canisters/Cs. |
| Saturation Solution | 1.56% Buffered Phenol, 0.06% Sodium Phenate, 98.38% Inert Ingredients |
| pH Range | 7.5 to 8.5 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction