Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contec™ Sporicidin™ Disinfectant Wipes
BIOPHARMA

Catalog No. 19159221
Click to view available options
Dimensions (L x W):
9.5 x 12 in.
5 x 8 in.
This item is not returnable. View return policy
This item is not returnable. View return policy

Clean, disinfect, and deodorize hard surfaces in critical areas. Contec™ Sporicidin™ Disinfectant Wipes use a broad spectrum disinfectant that provides 100% kill of disease and odor causing organisms. These wipes are alcohol and bleach-free.

  • Provides 100% kill of pathogenic vegetative organisms, including MRSA, VRE and Pandemic 2009 H1N1 Influenza A Virus (as well as H9N2)
  • Continuous residual activity up to 6 months after use
  • Wipes are non-staining, non-abrasive and non-corrosive
  • Recommended for use on treated wood, metal, painted surfaces, glass, porcelain, glazed tile, plastic, rubber, and vinyl materials
  • Provides users with a product safety rated in the lowest EPA toxicity categories
  • EPA and FDA registered; OSHA compliant
  • Non-hazardous disinfectant solution has no special shipping or disposal restrictions

Compliance

EPA registered. OSHA compliant.

Specifications

Product Type Sporicidin™ Disinfectant Wipes
Certifications/Compliance EPA, OSHA
Description Fresh Scent
Dimensions (L x W) 9.5 x 12 in.
For Use With (Application) Wiping cabinets, laminar flow hoods and other surfaces, compatible with Plastics, Wood, Glass and Metal, hard surfaces in critical areas
Packaging Quantity 85/Canister, 6 Canisters/Cs.
Saturation Solution 1.56% Buffered Phenol, 0.06% Sodium Phenate, 98.38% Inert Ingredients
pH Range 7.5 to 8.5
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.