Learn More
Contec™ Drip Caps for Multibucket Systems
BIOPHARMA
Catalog No. p-4422962 Shop All Contec Inc. Products
Click to view available options
For Use With (Equipment):
5 gal. multi-Bucket System
6.5 gal. multi-Bucket System
Includes
A stainless-steel connecting kit to prevent drips between the buckets.
Fits Contec™ Bucket Systems. The Contec™ Stainless Steel Drip Cap is easy to install and prevents drips between buckets.
  • Compatible with all disinfectant solutions and common solvents
  • 100% stainless steel for durability and cleanliness
  • Autoclavable
Specifications
Autoclavable Autoclavable
For Use With (Equipment) 6.5 gal. multi-Bucket System
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.