Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contec™ VertiKlean™ MAX™ Mop Heads
BIOPHARMAGREENER_CHOICE

Catalog No. 19169127 Shop All Contec Inc. Products
Click to view available options
Dimensions:
21 x 11 x 2 cm
24 x 13 x 1 cm
32 x 11 x 2 cm
38 x 11 x 2 cm
38 x 13 x 1 cm
46 x 13 x 1 cm
Sterility:
Non-sterile
Sterile
Material:
Hydrophilic Urethane Foam
Polyester, Foam
Ribbed Polyester Microfiber, Foam
Ribbed Polyester Microfiber, Hydrophilic Urethane Foam
Head Type:
Non-sealed Edge
Ribbed Sealed Edge
Sealed Edge

Clean walls, ceilings and other hard-to-reach surfaces with ease. VertiKlean™ MAX™ Mop Heads easily snap onto frames and last longer than traditional mops.

  • Features a single layer of hydrophilic polyurethane foam laminated to textured polyester microfiber fabric
  • Backed by a semi-rigid thermoformed plate that easily snaps onto mop head frame
  • Designed for applying disinfectants to both horizontal and vertical surfaces
  • Mop heads are composed of a disposable absorbent sponge laminated to filament polyester knit fabric and backed with a thermoformed plate that easily snaps onto VertiKlean mop hardware
  • Now available with ribbed microfiber for better contamination removal
  • Pivoting mop head frame is ideal for cleaning walls, floors, and ceilings as well as hard-to-reach areas such as fume hoods, mini environments, and around light fixtures and heavy equipment
  • Sterile mop heads are validated sterile per AAMI ISO11137 guidelines
  • Autoclaving mop heads is not recommended due to thermoformed plate; however, all hardware is autoclavable

Order Info

Frames, handles and bucket wringer system sold separately.

Certifications

Validated sterile to a Sterility Assurance Level of 10-6 per AAMI ISO11137:2006 Guidelines. Certificate of sterility is available.

Specifications

Dimensions 46 x 13 x 1 cm
Color Code White
Product Type Mop Head
Head Type Sealed Edge
Sterility Non-sterile
Autoclavable Not Autoclavable
Disposable Single-use
Quantity 4/bag; 5 bags/case
For Use With (Application) Wall Washing System
Color-coded No
Material Ribbed Polyester Microfiber, Hydrophilic Urethane Foam
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.