Learn More
Science First™ Contour Map Making Kit
SDPSCI_ED
Catalog No. p-2373532
Click to view available options
Quantity:
15
Each
Includes
  • Plastic sheet
  • Plastic mountain
  • Plastic box, 12L x 6W x 3 in.H
Learn how to read and make contour maps.
Easy to use—just cover the molded plastic model with water, mark mountain at each elevation, and then trace contour lines from above onto a transparent plastic sheet.
Specifications
Includes Plastic sheet, Plastic mountain, Plastic box
Product Type Contour Map Making Kit
Quantity Each
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.