Learn More
Siemens Healthineers Control Plasma
Catalog No. p-7235888 Shop All Siemens Healthineers Products
Click to view available options
Type:
Negative Control
Positive Control
This item is not returnable. View return policy
Catalog No. 23-044-683
Supplier: Siemens Healthineers 10446235
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Assayed control used to monitor the performance of coagulation assays in normal and pathological ranges.
  • Control Plasma P is citrated human plasma from selected healthy blood donors.
    • This control is an assayed control intended to monitor the performance of various analytes in the pathological range.
  • Control Plasma N is citrated normal human pooled plasma from selected healthy blood donors.
    • This control is an assayed control used to monitor the performance of various analytes in the normal range.
TRUSTED_SUSTAINABILITY
Specifications
Type Negative Control
For Use With (Application) Coagulation Testing
For Use With (Equipment) Coagulation Analyzers
Quantity 10 x 1 mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.