Learn More
Mopec™ Control Slides, Special Stains, Striations-Phosphotungstic Acid Hematoxylin Stain
Click to view available options
Includes:
10 Unstained, 1 Stained Slide
20 Unstained, 2 Stained Slides
98 Unstained, 2 Stained Slides
Description
- Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60°F to 80°F in a humidity controlled and light-deprived environment
- Case of 10 contains 10 unstained slides and 1 stained slide
- Case of 20 contains 20 unstained slides and 2 stained slides
- Case of 100 contains 98 unstained slides and 2 stained slides
Specifications
Specifications
Type | Control Slide |
Includes | 10 Unstained, 1 Stained Slide |