Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Controlled Environment Boot Cover

Catalog No. 191650132X Shop All Fisherbrand Products
Click to view available options
Size:
2X-Large
Universal
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Fisherbrand high top anti-skid boot covers fit over the calf with an elastic top to provide a comfortable fit and to prevent the boot cover from sliding down. Sole material is clean and durable. Upper material is soft and strong for all day comfort.

  • Inherent non-skid physical properties
  • Strong enough to withstand even the most abrasive floor surfaces
  • Fabric provides barrier protection and exhibits low particle generation
  • High COF fit

Specifications

Size 2X-Large
Material Polyethylene, Polypropylene
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Top and Ankles
Sole Type Anti-skid
Seam Type Serged
For Use With (Application) Controlled and Critical Environments
Packaging Quantity 200/Cs.
Packaging Type Double Bagged
Resistance Abrasion
Product Type Controlled Environments Boot Covers
Cleanroom Class Class 100 (ISO 5)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.