Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roche Diagnostics Controls for cobas c 111 Chemistry Analyzer

Catalog No. 22045820 Shop All Roche Diagnostics Products
Change view
Click to view available options
Type:
Ammonia/Ethanol/CO2 Normal Control
Precinorm U plus Control
Precipath Protein Control
Precipath U plus Control
Quantity:
3 x 1 mL
5 x 4 mL
10 x 3 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity
22-045-820 Precipath U plus Control 10 x 3 mL
22-045-821 Ammonia/Ethanol/CO2 Normal Control 5 x 4 mL
22-045-829 Precipath Protein Control 3 x 1 mL
22-045-819 Precinorm U plus Control 10 x 3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-045-820 Supplier Roche Diagnostics Supplier No. 12149443160
This item is not returnable. View return policy

Specifications

Format Bottle
For Use With (Equipment) Cobas™ c 111 chemistry analyzer
Type Precipath U plus Control
Quantity 10 x 3 mL
Form Lyophilized Serum, Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.