Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roche Diagnostics Controls for cobas c 111 Chemistry Analyzer

Catalog No. p-3592456
Click to view available options
Type:
Ammonia/Ethanol/CO2 Normal Control
Precinorm PUC Control
Precinorm U plus Control
Precipath PUC Control
Precipath Protein Control
Precipath U plus Control
Quantity:
3 x 1 mL
4 x 3 mL
5 x 4 mL
10 x 3 mL

Specifications

For Use With (Equipment) Cobas™ c 111 chemistry analyzer
Type Precipath PUC Control
Quantity 4 x 3 mL
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.