Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ CoolBox™ XT Systems
BIOPHARMA

Catalog No. 07210023 Shop All Corning Products
Change view
Click to view available options
Color:
Green
Orange
Pink
Purple
Includes:
Holder and One Cooling Core
Holder and Two Cooling Cores
Product Line:
CoolBox 2XT
CoolBox XT
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Includes Product Line
07-210-023 Orange Holder and One Cooling Core CoolBox XT
07-210-021 Purple Holder and One Cooling Core CoolBox XT
07-210-022 Green Holder and One Cooling Core CoolBox XT
07-210-024 Pink Holder and One Cooling Core CoolBox XT
07-210-025 Purple Holder and Two Cooling Cores CoolBox 2XT
07-210-026 Green Holder and Two Cooling Cores CoolBox 2XT
07-210-027 Orange Holder and Two Cooling Cores CoolBox 2XT
07-210-028 Pink Holder and Two Cooling Cores CoolBox 2XT
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 07-210-023 Supplier Corning™ Supplier No. 432023
Add to Cart
Add to Cart

Provide sample cooling or freezing without ice, electricity, or batteries.

Corning CoolBox XT systems are bench top cooling workstations that provide sample cooling or freezing without ice, electricity or batteries. CoolBox systems are versatile and accommodate a variety of sample formats and temperatures. The modular design enables the use of CoolRack™ and CoolSink™ thermo-conductive sample modules to hold microfuge tubes, cryogenic vials, PCR tubes or plates, assay plates and more.

  • Consists of a base, cooling cartridge/core and a lid
  • Includes a magnetized collar and a 1° to 8°C temperature indicator for visual assurance of proper temperature
  • To use with any 30 place Corning CoolRack Tubes Module
  • The internal cooling or freezing cores provide 0.5° to 4°C for over 10 hours with lid open and over 16 hours with lid closed
  • CoolBox systems may also be used with dry ice to create a compact, portable, snap freezing workstation for bacteria, virus or proteins.
TRUSTED_SUSTAINABILITY

Specifications

Description Benchtop cooling system
Color Orange
Type Cool Box
For Use With (Application) Enables Cooling for 16 Hours, Freezing for 8 Hours
Holds 30 Place
Includes Holder and One Cooling Core
Product Line CoolBox XT
Weight (Metric) 3 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.