Learn More
Cytiva Cooling Tube for Electrophoresis Units
Catalog No. 45001599 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
MultiTemp III Thermostatic Circulator Electrophoresis Unit
Catalog No. For Use With (Equipment)
45-001-599 MultiTemp III Thermostatic Circulator Electrophoresis Unit
1 options
Catalog No. 45-001-599 Supplier Cytiva Supplier No. 80110656
For use circulating coolant via the MultiTemp III Thermostatic Circulator to electrophoresis units such as the Multiphor II
  • Cooling tubing: 8mm i.d.,12 mm o.d. in 4 m pack size
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) MultiTemp III Thermostatic Circulator Electrophoresis Unit
Type Cooling Tube
Length (English) 157.48 in.
Length (Metric) 400 cm