Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ CoolRack™ M Microcentrifuge Tube Modules
BIOPHARMA

Catalog No. 07210037
Click to view available options
Holds:
15 x 1.5 or 2 mL MCT Tubes
24 x 1.5 or 2 mL MCT Tubes, SBS-compatible
30 x 1.5 or 2 mL MCT Tubes
6 x 1.5 or 2 mL MCT Tubes
90 x 1.5 or 2 mL MCT Tubes
96 x 1.5 or 2 mL MCT Tubes with A to H and 1 to 12
Color:
Gray
Green
Orange
Design:
M15
M24
M30
M6
M90
M96
This item is not returnable. View return policy
This item is not returnable. View return policy

Thermo-conductive microfuge tube modules that eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources.

Corning CoolRack thermo-conductive microcentrifuge tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.

  • Ensures +/- 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
TRUSTED_SUSTAINABILITY

Specifications

Color Gray
For Use With (Application) Cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen
For Use With (Equipment) Microcentrifuge Tubes
Holds 15 x 1.5 or 2 mL MCT Tubes
Array 3 x 5
Design M15
Type Microcentrifuge Tube Modules
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.