Learn More
Corning™ CoolRack™ XT PCR Tube Modules
BIOPHARMAGSA_VA
Catalog No. p-6830001
Click to view available options
Holds:
1.5 to 2 mL Tubes
384-well PCR Plate
96-well PCR Plate
This item is not returnable. View return policy
This item is not returnable. View return policy
Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
Corning CoolRack thermo-conductive tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to > 100°C and it will rapidly adapt to that temperature.
  • CoolRack modules ensure ± 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
  • SBS-compatible.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description Capacity: 96-well PCR microplate, 12 stripwells or microcentrifuge tubes, 1 SBS-compatible
Holds 96-well PCR Plate
Design PCR96
Type PCR Tube Module
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.