Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coombs+G193 Test, Polyclonal, 5mL, MP Biomedicals
Supplier: MP Biomedicals, Inc 08646351
Description

Specifications
Coomb's Test | |
Unconjugated | |
Primary |
Polyclonal | |
5 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction