Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COP Antibody, Novus Biologicals™
SDP

Catalog No. p-200041507 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB437416 25 μL
NBP230778 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB437416 Supplier Novus Biologicals Supplier No. NBP23077825UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

COP Polyclonal specifically detects COP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen COP
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q5EG05
Gene Alias CARD only domain-containing protein 1, CARD only protein, caspase recruitment domain family, member 16, Caspase-1 inhibitor COP, COP1caspase-1 dominant-negative inhibitor pseudo-ICE, COPcaspase recruitment domain-containing protein 16, EC 3.4.22.36, Pseudo interleukin-1 beta converting enzyme, pseudo interleukin-1beta converting enzyme, Pseudo-ICE
Gene Symbols CARD16
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: LHKGSLYPRIPGLGKELQTGTHKLS
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 114769
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.