Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COP9 Antibody (OTI3E1) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271382
Description
COP9 Monoclonal specifically detects COP9 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
COP9 | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000 | |
COP9 constitutive photomorphogenic homolog subunit 3 (Arabidopsis), COP9 signalosome complex subunit 3, CSN3COP9 complex subunit 3, JAB1-containing signalosome subunit 3, SGN3COP9 (constitutive photomorphogenic, Arabidopsis, homolog) subunit 3, Signalosome subunit 3 | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication, Neuronal Cell Markers, Neurotransmission, Signal Transduction | |
8533 | |
Human, Mouse, Rat | |
Lyophilized |
Western Blot | |
OTI3E1 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
COPS3 | |
Human recombinant protein fragment corresponding to amino acids 224-423 of human COPS3(NP_003644) produced in E.coli. | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction