Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COP9 signalosome complex subunit 2 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $426.00

Specifications

Antigen COP9 signalosome complex subunit 2
Concentration 1 mg/mL
Dilution Western Blot 1:100-1:2000
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB017910
SDP
View Documents
Novus Biologicals
NB10093557SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB10093557
SDP
View Documents
Novus Biologicals
NB10093557
0.1 mg
Each for $426.00
Only null left
Add to Cart
 
Description

Description

COP9 signalosome complex subunit 2 Polyclonal specifically detects COP9 signalosome complex subunit 2 in Human, Mouse, Rat, Drosophila samples. It is validated for Western Blot.
Specifications

Specifications

COP9 signalosome complex subunit 2
Western Blot 1:100-1:2000
Polyclonal
Rabbit
Cell Cycle and Replication, Neuronal Cell Markers, Neurotransmission, Signal Transduction
PBS with 0.02% Sodium Azide
9318
Synthetic peptide of residues 239-250 [ECGGKMHLREGE].
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1 mg/mL
Western Blot
Unconjugated
RUO
Drosophila, Mouse, Rat, Human
ALIEN, Alien homolog, COP9 constitutive photomorphogenic homolog subunit 2 (Arabidopsis), COP9 signalosome complex subunit 2, CSN2JAB1-containing signalosome subunit 2, SGN2TR-interacting protein 15, Signalosome subunit 2, thyroid receptor interacting protein 15, Thyroid receptor-interacting protein 15, TRIP15TRIP-15
COPS2
IgG
Affinity Purified
This product shares 100% homology to mouse, rat and Drosophila melanogaster alien isoform B protein.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.