Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COP9 signalosome complex subunit 2 Antibody, Novus Biologicals™
SDP

Catalog No. NB100363 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100363 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100363 Supplier Novus Biologicals Supplier No. NB100363
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

COP9 signalosome complex subunit 2 Polyclonal specifically detects COP9 signalosome complex subunit 2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen COP9 signalosome complex subunit 2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:1000 - 1:5000
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_004227.1
Gene Alias ALIEN, Alien homolog, COP9 constitutive photomorphogenic homolog subunit 2 (Arabidopsis), COP9 signalosome complex subunit 2, CSN2JAB1-containing signalosome subunit 2, SGN2TR-interacting protein 15, Signalosome subunit 2, thyroid receptor interacting protein 15, Thyroid receptor-interacting protein 15, TRIP15TRIP-15
Gene Symbols COPS2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human COP9 constitutive photomorphogenic homolog subunit 2 using the numbering given in entry NP_004227.1 (GeneID 9318).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Neuronal Cell Markers, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9318
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Rat, Zebrafish, X. laevis, X. tropicalis, Chicken, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Pig, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, and Chinese hamster.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.