Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COPA Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501567916
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-7916 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-7916 Supplier Bethyl Laboratories, Inc Supplier No. A304515AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine

Coatomer proteins are involved in regulating transport between the endoplasmic reticulum (ER) and the Golgi complex and in intra-Golgi transport. There exist two coatomer-protein mechanisms (COPI and COPII) and although they have mechanistic parallels, they are molecularly distinct. The COPI coat is comprised of seven subunits (alpha-, beta-, beta′-, gamma-, delta-, epsilon-, and zeta-COP) in a complex called coatomer. Assembly of the coatomer (COPI) onto non-clathrin coated vesicles is regulated by ADP-ribosylation factor (ARF). Vesicle formation, budding, fusion, and disassembly is dependent on GDP-GTP exchange, COPI, and ARF. COPI has been shown to facilitate retrograde intracellular transport from the ER to the Golgi complex. By contrast, COPII facilitates anterograde transport between these subcellular organelles. COPII has been shown to be independently and selectively recruited to the ER relative to COPI subunits.

Specifications

Antigen COPA
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene COPA
Gene Accession No. P53621, Q8CIE6
Gene Alias Coatomer subunit alpha, Alpha-coat protein, HEPCOP, HEP-COP, COPA
Gene Symbols Copa
Host Species Rabbit
Immunogen Between 1174 and 1224
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12847, 1314
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.