Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copan Diagnostics Calibrated Disposable Inoculating Loops and Needles in Zip Lock Pack
SDP

Catalog No. 1490631 Shop All Copan Diagnostics Inc. Products
Click to view available options
Product Type:
Inoculating Loop, Flexible
Inoculating Loop, Rigid
Inoculating Needle
Color:
Dark Blue
Dark Green
Light Blue
Light Green
Violet
This item is not returnable. View return policy
This item is not returnable. View return policy

Calibrated 1 and 10μL plastic disposable inoculating loops and needles

Calibrated 1 and 10μL plastic disposable inoculating loops and needles

Zip-lock closure reseals the pack between use to avoid contamination. Re-sealable bag allows the user to open and close the loop pack to remove individual loops.

  • Disposable, sterile plastic loops and needles manufactured from soft-flexible or hard-rigid plastic
  • Plastic injection molding technology ensures product consistency and eliminates variations in loop thickness and internal diameter
  • Ultrasmooth surfaces allow easy streaking without cutting or gouging agar surface
  • Calibrated using the Evans Blue Dye Method recommended by the American Society for Microbiology
  • Color coded for easy product identification
  • Packed in tamper-proof bags to guarantee sterility
  • Certificate of calibration included in each case

Specifications

Disposable Single-use
Length (English) 7.84 in.
Length (Metric) 20 cm
Diameter (Metric) Inside 0.14 cm
Material Plastic
Sterility Sterile
Diameter (English) Inside 0.057 in.
Color Violet
Product Type Inoculating Needle
Includes Evans blue dye certificate of calibration
Packaging Type Tamperproof Ziplock resealable bag
Calibrated Yes
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.