Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COPB2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1202915
Description
COPB2 Polyclonal specifically detects COPB2 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
COPB2 | |
Polyclonal | |
Unconjugated | |
PBS with 1 mg/ml BSA with 0.05% Sodium Azide | |
COPB2 | |
Synthetic peptide corresponding to residues K(494) V L A A Q E T H E G V T E D G I E(511) of rat beta-COP. | |
100 μg | |
Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience | |
9276 | |
Human, Rat | |
IgG |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
1 mg/mL | |
Western Blot 2 ug/ml, Flow Cytometry 3-5 ug/10^6 cells, Immunocytochemistry/Immunofluorescence 2 ug/ml | |
beta prime subunit, Beta'-coat protein, Beta'-COP, betaprime-COP, coatomer protein complex, subunit beta 2 (beta prime), coatomer subunit beta', p102 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Detects coatomer-protein I beta (COP I beta). | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction