Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COPB2 Recombinant Rabbit Monoclonal Antibody (1X5F7)
GREENER_CHOICE

Catalog No. FEMA551300
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551300 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA551300 Supplier Invitrogen™ Supplier No. MA551300
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: AYQLAVEAES EQKWKQLAEL AISKCQFGLA QECLHHAQDY GGLLLLATAS GNANMVNKLA EGAERDGKNN VAFMSYFLQG KVDACLELLI RTGRLPEAAF LARTYLPSQV SRVVKLWREN LSKVNQKAAE SLADPTEYEN LFPGLKEAFV VEEWVKETHA DLWPAKQYPL VTPNEERNVM EEGKDFQPSR STAQQELDGK PASPTPVIVA SHTANKEEKS LLELEVDLDN LELEDIDTTD INLDEDILDD.

The coatomer is a cytosolic protein complex that binds to dilysine motifs and reversibly associates with Golgi non- clathrin-coated vesicles, which further mediate biosynthetic protein transport from the ER, via the Golgi up to the trans Golgi network. Coatomer complex is required for budding from Golgi membranes, and is essential for the retrograde Golgi-to-ER transport of dilysine-tagged proteins. In mammals, the coatomer can only be recruited by membranes associated to ADP-ribosylation factors (ARFs), which are small GTP-binding proteins; the complex also influences the Golgi structural integrity, as well as the processing, activity, and endocytic recycling of LDL receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COPB2
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 1X5F7
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene COPB2
Gene Accession No. O55029, P35606
Gene Alias AI256832; beta prime COP; beta'-coat protein; Beta'-COP; betaprime-COP; coatomer binding complex, beta prime subunit; coatomer protein complex subunit beta 2; coatomer protein complex subunit beta 2 (beta prime); coatomer protein complex subunit beta prime; coatomer protein complex, subunit beta 2 (beta prime); coatomer subunit beta'; COPB2; p102
Gene Symbols COPB2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 657-906 of human COPB2 (NP_004757.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50797, 9276
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.