Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COPD Monoclonal Antibody (P189)

Catalog No. PIMA518294
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518294 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518294 Supplier Invitrogen™ Supplier No. MA518294
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Recommended positive controls: 293T, A431, HeLa, HepG2, Neuro 2A, GL261, C8D30, NIH-3T3, BCL-1, Raw264.7, C2C12, PC-12, Rat2. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (82%), Chicken (92%), Rhesus Monkey (99%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene maps in a region, which include the mixed lineage leukemia and Friend leukemia virus integration 1 genes, where multiple disease-associated chromosome translocations occur. It is an intracellular protein. Archain sequences are well conserved among eukaryotes and this protein may play a fundamental role in eukaryotic cell biology. It has similarities to heat shock proteins and clathrin-associated proteins, and may be involved in vesicle structure or trafficking.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COPD
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone P189
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ARCN1
Gene Accession No. P48444, Q5XJY5, Q66H80
Gene Alias 4632432M07Rik; Archain; archain 1; archain vesicle transport protein 1; Arcn1; coatomer delta subunit; coatomer delta-COP; coatomer protein complex, subunit delta; coatomer protein delta-COP; coatomer subunit delta; COPD; delta-coat protein; Delta-COP; nur17; pale coat neuro
Gene Symbols ARCN1
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human COPD. The exact sequence is proprietary.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 213827, 300674, 372
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.