Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COPE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560425
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560425 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560425 Supplier Invitrogen™ Supplier No. PA560425
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GSGEVDELFD VKNAFYIGSY QQCINEAQRV KLSSPERDVE RDVFLYRAYL AQRKFGVVLD EIKPSSAPEL QAVRMFADYL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.

The product of this gene is an epsilon subunit of coatomer protein complex. Coatomer is a cytosolic protein complex that binds to dilysine motifs and reversibly associates with Golgi non-clathrin-coated vesicles. It is required for budding from Golgi membranes, and is essential for the retrograde Golgi-to-ER transport of dilysine-tagged proteins. Coatomer complex consists of at least the alpha, beta, beta', gamma, delta, epsilon and zeta subunits. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COPE
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COPE
Gene Accession No. O14579
Gene Alias 1110005D17Rik; coatomer epsilon subunit; coatomer protein complex subunit epsilon; coatomer protein complex, subunit epsilon; coatomer subunit epsilon; Cope; Cope1; epsilon coat protein; epsilon-coat protein; Epsilon-COP; nonclathrin coat protein epsilon-COP
Gene Symbols COPE
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COPE. Recombinant protein control fragment (Product #RP-102946).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11316
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.