Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COPG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558146
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558146 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558146 Supplier Invitrogen™ Supplier No. PA558146
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VKQPEKVAAT RQEIFQEQLA AVPEFRGLGP LFKSSPEPVA LTESETEYVI RCTKHTFTNH MVFQFDCTNT LNDQTLENVT VQMEPTE Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

The coatomer is a cytosolic protein complex that binds to dilysine motifs and reversibly associates with Golgi non-clathrin-coated vesicles, which further mediate biosynthetic protein transport from the ER, via the Golgi up to the trans Golgi network. Coatomer complex is required for budding from Golgi membranes, and is essential for the retrograde Golgi-to-ER transport of dilysine-tagged proteins. In mammals, the coatomer can only be recruited by membranes associated to ADP-ribosylation factors (ARFs), which are small GTP-binding proteins; the complex also influences the Golgi structural integrity, as well as the processing, activity, and endocytic recycling of LDL receptors. Required for limiting lipid storage in lipid droplets. Involved in lipid homeostasis by regulating the presence of perilipin family members PLIN2 and PLIN3 at the lipid droplet surface and promoting the association of adipocyte triglyceride lipase (PNPLA2) with the lipid droplet surface to mediate lipolysis. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen COPG
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Copg1
Gene Accession No. Q4AEF8, Q9QZE5, Q9Y678
Gene Alias AU019265; BC056168; coat protein gamma-cop; coatomer protein complex subunit gamma 1; coatomer protein complex, subunit gamma 1; coatomer subunit gamma; coatomer subunit gamma-1; COPG; COPG1; D6Ertd71e; D6Wsu16e; Gamma-1-coat protein; gamma-1-COP; gamma-coat protein; gamma-COP
Gene Symbols Copg1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COPG. Recombinant protein control fragment (Product #RP-96463).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22820, 297428, 54161
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.