Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COPG2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100448
Description
Antibody detects endogenous levels of total COPG2.
The coatomer is a cytosolic protein complex that binds to dilysine motifs and reversibly associates with Golgi non-clathrin-coated vesicles, which further mediate biosynthetic protein transport from the ER, via the Golgi up to the trans Golgi network.
Specifications
COPG2 | |
Polyclonal | |
Unconjugated | |
Copg2 | |
2-COP; AW227625; cb943; coat protein, nonclathrin, gamma-2-cop; coatomer protein complex subunit gamma 2; coatomer protein complex, subunit gamma 2; coatomer protein gamma2-COP; coatomer subunit gamma-2; copg; Copg2; copg2 protein; COPI coat complex subunit gamma 2; fi31b06; FLJ11781; gamma-2-coat protein; Gamma-2-COP; gamma2-cop; gamma-cop; LOC101789299; RGD1566215; si:ch211-285d14.8; testicular secretory protein Li 12; wu:fi31b06 | |
Rabbit | |
Affinity chromatography | |
RUO | |
26958, 54160 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9QXK3, Q9UBF2 | |
Copg2 | |
A synthesized peptide derived from human COPG2(Accession Q9UBF2), corresponding to amino acid residues P714-V764. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction