Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COPG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590730
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590730 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590730 Supplier Invitrogen™ Supplier No. PA590730
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LNATYIFNGL TVSVPGMEKA LHQYTLEPSE KPFDMKSIPL AMAPVFEQKA EITLVATKPE KLAPSRQDIF QEQLAAIPEF LNIGPLFKSS EPVQLTEAET EYFVRCIKHM FTNHIVFQFD CTNTLNDQLL EKVTVQMEPS DSYEVLSCIP APSLPYNQPG ICYTLVRLPD DDPTAVAGSF SCTMKFTVRD CDPNTGVPDE DGYDDEYVLE DLEVTVSDHI QKVLKPNFAA AWEEVGDTFE KEETFALSST KTLEEAVNNI ITFLGMQPCE RSDKVPENKN SHSLYLAGIF RGGYDLLVRS RLALADGVTM QVTVRSKERT PVDVILASVG; Positive Samples: Mouse heart, Mouse kidney, Rat heart; Cellular Location: COPI-coated vesicle membrane, Cytoplasm, Cytoplasmic side, Cytoplasmic vesicle, Golgi apparatus membrane, Peripheral membrane protein, cytosol.

The coatomer is a cytosolic protein complex that binds to dilysine motifs and reversibly associates with Golgi non-clathrin-coated vesicles, which further mediate biosynthetic protein transport from the ER, via the Golgi up to the trans Golgi network.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COPG2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Copg2
Gene Accession No. D4ABY2, Q9QXK3, Q9UBF2
Gene Alias 2-COP; AW227625; cb943; coat protein, nonclathrin, gamma-2-cop; coatomer protein complex subunit gamma 2; coatomer protein complex, subunit gamma 2; coatomer protein gamma2-COP; coatomer subunit gamma-2; copg; Copg2; copg2 protein; COPI coat complex subunit gamma 2; fi31b06; FLJ11781; gamma-2-coat protein; Gamma-2-COP; gamma2-cop; gamma-cop; LOC101789299; RGD1566215; si:ch211-285d14.8; testicular secretory protein Li 12; wu:fi31b06
Gene Symbols Copg2
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26958, 301742, 54160
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.