Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper AA Standard, 1000 ppm Cu in 3% HNO3, Ricca Chemical
SDP

Catalog No. p-3251570 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7440-50-8, 7697-37-2, 7732-18-5
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Cu
CAS Min % 93.0, 4.46, 0.09
CAS Max % 97.0, 4.64, 0.1
Equivalence 1 mL = 1 mg Cu
Physical Form Liquid
Color Blue
pH < 2
Melting Point Approximately 0°C
Quantity 100 mL
Chemical Name or Material Copper AA Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.