Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Beryllium rod, alloy C17200, 4.76mm (0.188 in.) dia., Thermo Scientific™

Catalog No. AA42481BU
Change view
Click to view available options
Quantity:
25 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42481BU 25 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA42481BU Supplier Thermo Scientific Chemicals Supplier No. 42481BU
Only null left
Add to Cart

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Copper Beryllium rod, Alloy C17200
Name Note 4.76mm (0.188 in.) dia.
Avg. Mol. Wt. or Mol. Wt. Range Cu:Be; 98.1:1.9 wt%
Quantity 25 cm
Odor Odorless
Weight ≈37.3g/25cm
Form Rod
MDL Number MFCD01320550
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.