Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper chromite, catalyst, Thermo Scientific™

Catalog No. AC197821000
Click to view available options
:
100g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 12018-10-9
Quantity 100g
Molecular Formula Cr2CuO4
Linear Formula 2CuO·Cr2O3
MDL Number MFCD00064661
Fieser 01,156; 02,82
Merck Index 15, 2625
Synonym oxocopper; oxo-oxochromiooxy chromium, cucr2o4, cr2o3.cuo, copper ii chromium iii oxide, chromia; copper ii oxide, copper chromite, barium promoted cr2cuo cuobao graphite cro cr2o3
InChI Key FULFYAFFAGNFJM-UHFFFAOYSA-N
SMILES O=[Cr]O[Cr]=O.O=[Cu]
IUPAC Name oxocopper;oxo(oxochromiooxy)chromium
Molecular Weight (g/mol) 231.534
PubChem CID 11770311
Formula Weight 311.07
Percent Purity ≥36% (Cu) (on dry substance), ≥26% (Cr) (on dry substance)
Loss on Drying 0.5% max.
Packaging Plastic bottle
Chemical Name or Material Copper chromite, Catalyst
Show More Show Less
missing translation for 'healthHazard1' GHS Signal Word: Warning
missing translation for 'healthHazard2' GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
missing translation for 'healthHazard3' GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.