Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper etchant

Catalog No. p-4408710
Click to view available options
Quantity:
1 qt.
1 gal.

Iron (III) chloride solution for immersion etching copper, copper alloys and Kovar in printed circuit board applications

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Copper etchant
Density 1.19
Quantity 1 qt.
Form Liquid
MDL Number MFCD00161912
What is the etch rate of Copper etchant at 40 degrees C?

Copper etchant has an etch rate of 1 mil/minute at 40 degrees C (etches 1 oz copper in 90 seconds).

How do I reduce undercutting of an etchant?

Undercutting of the etchant can be reduced by increasing the rate of stirring or agitation.

Do I need to dilute an etchant?

No, it is a ready-to-use solution. Dilution will only be necessary if you want to reduce the etch rate.

How do I reduce the etch rate of an etchant?

The etch rate of the etchant can be reduced by adding 1 part deionized water to 2 parts etchant. This will reduce the etch rate approximately 50%.

How do I increase the etch rate of an etchant?

The etch rate of the etchant will be increased by increasing the rate of stirring or agitation.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.