Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Gluconate, FCC, 98-102%, Spectrum™ Chemical
SDP

Catalog No. p-7157151
Click to view available options
Quantity:
125 g
Packaging:
Poly Bottle

C1329, 527-09-3, C12H22CuO14

Spectrum™ Chemical Copper Gluconate, FCC is used as a nutrient and dietary supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 527-09-3
Molecular Formula C12H22CuO14
Molecular Weight (g/mol) 453.84
InChI Key OCUCCJIRFHNWBP-IYEMJOQQSA-L
IUPAC Name copper(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
SMILES [Cu++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O

Specifications

CAS Max % 100%
Quantity 125 g
Percent Purity 98 to 102%
Grade FCC
Packaging Poly Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.