Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(I) bromide, 99%, Thermo Scientific™

Catalog No. aa4721130
Click to view available options
Quantity:
250 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-70-4
Molecular Formula BrCu
Molecular Weight (g/mol) 143.45
MDL Number MFCD00010969
InChI Key NKNDPYCGAZPOFS-UHFFFAOYSA-M
Synonym copper i bromide, copper bromide cubr, copper 1+ bromide, copper monobromide, cuprousbromide, hsdb 270, cubr, copper 1 bromide
PubChem CID 24593
IUPAC Name λ¹-copper(1+) bromide
SMILES [Cu+].[Br-]

Specifications

Boiling Point 1345°C
Melting Point 492°C
Physical Form Powder
Quantity 250 g
UN Number UN3077
Merck Index 14,2659
Formula Weight 143.45
Percent Purity 99%
Sensitivity Air Sensitive
Density 4.98
Chemical Name or Material Copper(I) bromide
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement H315-H319-H335; Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 232-131-6
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.