Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amberlyst™ A-21, ion-exchange resin

Catalog No. AC202170010
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle

CAS: 9049-93-8

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ion exchange resin
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9049-93-8
MDL Number MFCD00145842

Specifications

Physical Form Beads
Particle Size Particle size analysis: uniformity coefficient: <1.80, harmonic mean size: 0.490 to 0.690mm,% <0.300mm: <1.0%,%>1.180mm: <2.0%
Packaging Plastic bottle
Volume Total Capacity (free base):> 1.30 eq/L
Water Retention Capacity (free base): 52.0 to 62.0%
Color Brown
Fieser 06,302; 07,182; 08,263
Merck Index 10, 381
Quantity 1 kg
Chemical Name or Material Amberlyst™ A-21

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.