Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(I) Chloride, Solstice

Catalog No. 6002089 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-020-89 25 g
60-020-90 2 kg
60-020-91 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-020-89 Supplier Solstice Supplier No. 21294625G

Reagent Grade, 97%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-89-6
Molecular Formula ClCu
Molecular Weight (g/mol) 98.996
MDL Number MFCD00010971
InChI Key OXBLHERUFWYNTN-UHFFFAOYSA-M
Synonym cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech
PubChem CID 62652
ChEBI CHEBI:53472
IUPAC Name chlorocopper
SMILES Cl[Cu]

Specifications

Quantity 25 g
Linear Formula CuCl
UN Number 2802
Solubility Information Partially soluble in dimethyl sulfide, Insoluble in water
Formula Weight 99g/mol
Percent Purity 97%
Grade Reagent
Vapor Pressure 1.3mmHg (546°C)
Chemical Name or Material Copper(I) Chloride
Hazard Statement P273-P280-P305 + P351 + P338-P501
Precautionary Statement H302-H315-H318-H410
EINECSNumber 231-842-9
RTECSNumber GL6990000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.