Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calcium oxide, 96%, extra pure
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
Specifications
Specifications
| Boiling Point | 2850.0°C |
| Melting Point | 2570.0°C |
| Color | Beige |
| Physical Form | Powder |
| Quantity | 1 kg |
| Assay Percent Range | 95% min. (Complexometry) |
| Linear Formula | CaO |
| Merck Index | 15, 1688 |
| Solubility Information | Solubility in water: reacts. Other solubilities: soluble in acids (reacts), soluble in glycerol, insoluble in alcohol |
| Formula Weight | 56.08 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye damage. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if pre |
| EINECSNumber | 215-138-9 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction