Learn More
Sodium Borohydride, 99%, powder

Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications
Specifications
Melting Point | 360°C |
Color | White |
Physical Form | Powder |
Quantity | 100 g |
Assay Percent Range | 99% |
Linear Formula | NaBH4 |
UN Number | UN 1426 |
Merck Index | 15, 8727 |
Formula Weight | 37.83 |
Percent Purity | ≥98.5% |
Show More |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS P Statement Causes severe skin burns and eye damage. Toxic if swallowed. May damage fertility. May damage the unborn child. In contact with water releases flammable gases which may ignite spontaneously. Reacts violently with water. |
Precautionary Statement | GHS P Statement Immediately call a POISON CENTER or doctor/physician. Keep away from any possible contact with water, because of violent reaction and possible flash fire. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection. |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.