Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(I) sulfide, 99.5% (metals basis)

Catalog No. p-3256998
Click to view available options
Quantity:
50 g
250 g
1 kg

For Methods: Luminous Paints, Electrodes, Solar Cells | CAS: 22205-45-4 | Cu2S | 159.15 g/mol

Copper(I) sulfide is used in luminous paints, electrodes and in solar cells. It is also used for grain mounts, XRD and in microprobe standards for the identification of unknown minerals. It acts as a precursor for the preparation of copper(I) oxide and copper metal. Further, it acts as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22205-45-4
Molecular Formula Cu2S
Molecular Weight (g/mol) 159.15
MDL Number MFCD00016062
InChI Key AQMRBJNRFUQADD-UHFFFAOYSA-N
Synonym copper i sulfide, cuprasulfide, copper sulfide cu2s, dicopper sulfide, dicopper sulphide, dicopper monosulfide, cuprous sulfide cu2s, unii-349m3c1rs1, cu2s, cupriosulfanyl copper
PubChem CID 62755
IUPAC Name bis(λ¹-copper(1+)) sulfanediide
SMILES [S--].[Cu+].[Cu+]

Specifications

Melting Point ∽1100°C
Color Black
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.5% (Metal basis)
Solubility Information Slightly soluble in ammonium hydroxide. Insoluble in water and acetic acid.
Formula Weight 159.14
Odor Odorless
Appearance Black powder
Density 5.6 g/cm3
Chemical Name or Material Copper(I) sulfide
Show More Show Less
EINECSNumber 244-842-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.