Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Copper(I) thiocyanate

Catalog No. 298212250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
50 g
250 g

Catalog No. 29-821-2250G

Supplier: Sigma Aldrich 298212250G

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The crystal belongs to the orthorhombic system, space group: Pbca,a=10.994(9)+a, b=7.224(3)+a,c=6.662(2)+a.The product can be prepared from Cu(CH3COO)2-+H2O, AgNO3, NH4NCS and im(im = imidazole). The structure and optical properties were reported. It is an air- and light-stable source of Cu(I).

Specifications

CAS 1111-67-7
Molecular Formula CCuNS
Linear Formula CuSCN
MDL Number MFCD00010980
Quantity 250 g
Synonym Cuprous thiocyanate
Molecular Weight (g/mol) 121.63
Percent Purity 99%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.