Learn More
Copper(II) acetate, anhydrous, Alfa Aesar™
Manufacturer:  Alfa Aesar 1941714
Quantity 25g
 View more versions of this product
Catalog No. AA1941714
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
142-71-2
Cu(OOCCH3)2
MFCD00008690
Powder
181.63
Copper(II) acetate, anhydrous
Specifications
142-71-2
Cu(OOCCH3)2
181.63
Odorless
25g
Powder
Green crystals
MFCD00008690
Green
Copper(II) acetate, anhydrous
Safety and Handling
SDS & Safety Documents
H318-H400-H410-H302; Causes serious eye damage. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed.
DOTInformation : DOT Class: 9, Packaging Group: III
EINECSNumber : 205-553-3