Learn More
Copper(II) acetate monohydrate, ACS, 98.0-102.0%
Description
Copper(II) acetate monohydrate is used in DNA extraction in biochemical applications. It is also used as a catalyst or oxidizing agent in organic syntheses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 115°C |
Color | Blue-Green |
Physical Form | Powder |
Quantity | 2 kg |
Assay Percent Range | 98.0-102.0% |
Solubility Information | Soluble in water and alcohol. Slightly soluble in ether and glycerin. |
Formula Weight | 199.65 (181.64 Anhydrous) |
Grade | ACS |
Odor | Odorless |
Appearance | Blue-green powder |
Show More |
Safety and Handling
Hazard Category | H302-H314-H335-H373 |
Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
DOTInformation | DOT Class: 8; Packaging Group: II |
EINECSNumber | 205-553-3 |
RTECSNumber | AG3500000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.