Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) acetate monohydrate, ACS, 98.0-102.0%

Catalog No. AA1417736
Click to view available options
Quantity:
100 g
500 g
2 kg

CAS: 6046-93-1 | C4H8CuO5 | 199.65 g/mol

Copper(II) acetate monohydrate is used in DNA extraction in biochemical applications. It is also used as a catalyst or oxidizing agent in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6046-93-1
Molecular Formula C4H8CuO5
Molecular Weight (g/mol) 199.65
MDL Number MFCD00149570
InChI Key NWFNSTOSIVLCJA-UHFFFAOYSA-L
Synonym copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate
PubChem CID 165397
IUPAC Name copper;diacetate;hydrate
SMILES O.[Cu++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 115°C
Color Blue-Green
Physical Form Powder
Quantity 500 g
Assay Percent Range 98.0-102.0%
Solubility Information Soluble in water and alcohol. Slightly soluble in ether and glycerin.
Formula Weight 199.65 (181.64 Anhydrous)
Grade ACS
Density 1.882
Chemical Name or Material Copper(II) acetate, monohydrate
Hazard Category H302-H314-H335-H373
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation DOT Class: 8; Packaging Group: II
EINECSNumber 205-553-3
RTECSNumber AG3500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.